Learn More
Waters Corp U(H)PLC Column, ACQUITY BEH HILIC, 1.7 μm, 130 Å, 2.1 x 50 mm
Catalog No. 50782158 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Acquity UPLC column is designed for high-performance liquid chromatography applications. It features a 50 mm length, 2.1 mm inner diameter, and a 1.7 μm particle size, making it suitable for various analytical separations.
  • Acquity column description
  • Hybrid package specification
  • 2.1 mm inner diameter
  • HILIC mode
  • 1.7 μm particle size
  • 130 Å pore size
  • L3 USP number
  • 50 mm length
Catalog No. 50-782-158 Supplier Waters Corp Supplier No. 186004053
This Acquity UPLC column is designed for high-performance liquid chromatography applications. It features a 50 mm length, 2.1 mm inner diameter, and a 1.7 μm particle size, making it suitable for various analytical separations.
  • Acquity column description
  • Hybrid package specification
  • 2.1 mm inner diameter
  • HILIC mode
  • 1.7 μm particle size
  • 130 Å pore size
  • L3 USP number
  • 50 mm length