Learn More
Waters Corp Waters Corporation ACQUITY UPLC BEH HILIC Column, 130Å, 1.7μm, 2.1 mm × 100 mm
Catalog No. 50781793 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. They are more rugged than silica-based HILIC phases, and the BEH particle's chemical stability results in long column lifetimes.
  • Designed to retain and separate very polar compounds.
  • Rugged BEH particle chemistry ensures long column lifetimes.
  • Improves retention for very polar basic analytes.
  • Offers exceptional chemical stability and peak shape.
  • Incorporates sub-2 μm particle technology.
  • Operates up to 15,000 psi (1000 Bar).
  • Enhances resolution, sample throughput, and sensitivity.
  • Can be used with VanGuard Pre-column for extended column lifetimes.
  • Batch tested chromatographically for excellent performance.
Catalog No. 50-781-793 Supplier Waters Corp Supplier No. 186003461
ACQUITY BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. They are more rugged than silica-based HILIC phases, and the BEH particle's chemical stability results in long column lifetimes.
  • Designed to retain and separate very polar compounds.
  • Rugged BEH particle chemistry ensures long column lifetimes.
  • Improves retention for very polar basic analytes.
  • Offers exceptional chemical stability and peak shape.
  • Incorporates sub-2 μm particle technology.
  • Operates up to 15,000 psi (1000 Bar).
  • Enhances resolution, sample throughput, and sensitivity.
  • Can be used with VanGuard Pre-column for extended column lifetimes.
  • Batch tested chromatographically for excellent performance.