Learn More
Waters Corp ACQUITY UPLC Column, HILIC, 1.7 μm, 130 Å, 3x50 mm
Catalog No. 50791184 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY BEH HILIC columns are designed for the efficient retention and separation of very polar compounds, optimized for reproducible HPLC HILIC separations. Their unbonded BEH substrate ensures enhanced ruggedness and chemical stability for extended column lifetimes.
  • Efficient retention and separation of very polar compounds.
  • Optimized for reproducible HPLC HILIC separations.
  • Enhanced ruggedness and extended column lifetimes.
  • Orthogonal separation mode for polar and ionizable chemical mixtures.
  • Wide pH stability (1–9) for extremely polar analytes.
  • Supports working pressures up to 18,000 psi (1240 bar).
  • Manufactured in a cGMP, ISO 9001 certified facility.
Catalog No. 50-791-184 Supplier Waters Corp Supplier No. 186004675
ACQUITY BEH HILIC columns are designed for the efficient retention and separation of very polar compounds, optimized for reproducible HPLC HILIC separations. Their unbonded BEH substrate ensures enhanced ruggedness and chemical stability for extended column lifetimes.
  • Efficient retention and separation of very polar compounds.
  • Optimized for reproducible HPLC HILIC separations.
  • Enhanced ruggedness and extended column lifetimes.
  • Orthogonal separation mode for polar and ionizable chemical mixtures.
  • Wide pH stability (1–9) for extremely polar analytes.
  • Supports working pressures up to 18,000 psi (1240 bar).
  • Manufactured in a cGMP, ISO 9001 certified facility.