Learn More
Waters Corp Waters ACQUITY UPLC HILIC Column, 130Å, 1.7 μm, 3x75 mm
Supplier:  Waters Corp 186005613
Encompass_Preferred
BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
  • Contain unbonded BEH substrate.
  • Designed to retain and separate very polar compounds.
  • Optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica based HILIC phases.
  • Chemical stability results in long column lifetimes.
Catalog No. 50-466-149