Learn More
Waters Corp Waters ACQUITY UPLC BEH HILIC Column, 130Å, 1.7 μm, 2.1 mm X 50 mm
Catalog No. 50781792 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These HILIC columns are designed for the retention and separation of very polar compounds, especially when reversed-phase chromatography is unsuitable. They feature rugged BEH particle technology for enhanced chemical stability and extended column lifetimes compared to silica-based HILIC phases, delivering efficient and reproducible separations for UPLC systems.
  • Retains and separates very polar compounds.
  • More rugged than silica-based HILIC phases due to BEH particle’s chemical stability.
  • Offers wide usable pH range for improved stability and extended column lifetimes.
  • Optimized for maximum chromatographic performance with UPLC systems.
  • Chromatographically tested to ensure superior performance.
  • Includes Performance Chromatogram and Certificate of Batch Analysis.
  • Features eCord Intelligent Chip Technology for lifetime performance history and digital quality control.
Catalog No. 50-781-792 Supplier Waters Corp Supplier No. 186003460
These HILIC columns are designed for the retention and separation of very polar compounds, especially when reversed-phase chromatography is unsuitable. They feature rugged BEH particle technology for enhanced chemical stability and extended column lifetimes compared to silica-based HILIC phases, delivering efficient and reproducible separations for UPLC systems.
  • Retains and separates very polar compounds.
  • More rugged than silica-based HILIC phases due to BEH particle’s chemical stability.
  • Offers wide usable pH range for improved stability and extended column lifetimes.
  • Optimized for maximum chromatographic performance with UPLC systems.
  • Chromatographically tested to ensure superior performance.
  • Includes Performance Chromatogram and Certificate of Batch Analysis.
  • Features eCord Intelligent Chip Technology for lifetime performance history and digital quality control.