Learn More
Waters Corp Waters Corporation ACQUITY UPLC BEH HILIC Column, 1.7 μm, 3.0 x 50 mm, 130Å
Catalog No. 50791043 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC BEH HILIC Column is designed for high-speed liquid chromatography applications. It features a 1.7 μm particle size, 3.0 x 50 mm dimensions, and 130Å pore size, offering improved retention for very polar species and robust, reproducible separations. This column is optimized for UPLC HILIC conditions and provides an orthogonal separation mode for complex mixtures of polar and ionizable compounds. Each pack contains 3 columns.
  • Improved retention of very polar species.
  • Provides an orthogonal separation mode for mixtures of polar and ionizable compounds.
  • Optimized and tested for efficient, reproducible separations under UPLC HILIC conditions.
  • Designed to overcome chemical stability issues common in HILIC stationary phases.
  • Rugged BEH particle with a wide usable pH range, leading to long column lifetimes.
Catalog No. 50-791-043 Supplier Waters Corp Supplier No. 176001810
Add to Cart
Add to Cart
The ACQUITY UPLC BEH HILIC Column is designed for high-speed liquid chromatography applications. It features a 1.7 μm particle size, 3.0 x 50 mm dimensions, and 130Å pore size, offering improved retention for very polar species and robust, reproducible separations. This column is optimized for UPLC HILIC conditions and provides an orthogonal separation mode for complex mixtures of polar and ionizable compounds. Each pack contains 3 columns.
  • Improved retention of very polar species.
  • Provides an orthogonal separation mode for mixtures of polar and ionizable compounds.
  • Optimized and tested for efficient, reproducible separations under UPLC HILIC conditions.
  • Designed to overcome chemical stability issues common in HILIC stationary phases.
  • Rugged BEH particle with a wide usable pH range, leading to long column lifetimes.