Learn More
Waters Corp Waters ACQUITY UPLC BEH HILIC Column, 130Å, 1.7 μm, 1x50 mm
Catalog No. 50781789 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These HILIC columns with unbonded BEH substrate retain and separate very polar compounds. Optimized for efficient, reproducible HPLC HILIC separations, they are more rugged than silica-based HILIC phases, offering long column lifetimes due to BEH particle chemical stability.
  • HILIC chemistry
  • Hydrophilic interaction separation
  • Hybrid particle substrate
  • pH range 1 to 9
  • Maximum pressure of 18000 psi (1240 Bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • Particle size of 1.7 μm
  • Parker-style endfitting
  • Pore size of 130 Å
  • Column format
  • Surface area of 185
  • UPLC and UHPLC system compatible
  • BEH particle technology
  • L3 USP classification
  • 1 mm inner diameter
  • 50 mm length
  • eCord enabled
  • 1 per package
Catalog No. 50-781-789 Supplier Waters Corp Supplier No. 186003457
These HILIC columns with unbonded BEH substrate retain and separate very polar compounds. Optimized for efficient, reproducible HPLC HILIC separations, they are more rugged than silica-based HILIC phases, offering long column lifetimes due to BEH particle chemical stability.
  • HILIC chemistry
  • Hydrophilic interaction separation
  • Hybrid particle substrate
  • pH range 1 to 9
  • Maximum pressure of 18000 psi (1240 Bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • Particle size of 1.7 μm
  • Parker-style endfitting
  • Pore size of 130 Å
  • Column format
  • Surface area of 185
  • UPLC and UHPLC system compatible
  • BEH particle technology
  • L3 USP classification
  • 1 mm inner diameter
  • 50 mm length
  • eCord enabled
  • 1 per package