Learn More
Waters Corp Waters ACQUITY UPLC BEH HILIC Column, 130Å, 1.7 μm, 2.1 x 100 mm
Catalog No. 50780031 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY BEH HILIC columns are designed for retaining and separating very polar compounds via Hydrophilic-Interaction Chromatography (HILIC). They feature rugged 1.7 μm unbonded BEH particles for efficient, reproducible UPLC HILIC separations, offering superior chemical stability and an extended usable pH range for long column lifetimes.
  • Retains and separates very polar compounds.
  • Optimized for efficient, reproducible HPLC HILIC separations.
  • Rugged BEH particles offer superior chemical stability and extended column lifetimes.
  • Wide usable pH range addresses silica-based HILIC instability.
  • Packing materials are purpose-built for UPLC systems.
  • Manufactured in cGMP, ISO 9001 certified facilities with ultrapure reagents.
  • Each batch undergoes rigorous chromatographic testing.
  • Individually tested; Performance Chromatogram and Certificate of Batch Analysis included.
Catalog No. 50-780-031 Supplier Waters Corp Supplier No. 176001093
The ACQUITY BEH HILIC columns are designed for retaining and separating very polar compounds via Hydrophilic-Interaction Chromatography (HILIC). They feature rugged 1.7 μm unbonded BEH particles for efficient, reproducible UPLC HILIC separations, offering superior chemical stability and an extended usable pH range for long column lifetimes.
  • Retains and separates very polar compounds.
  • Optimized for efficient, reproducible HPLC HILIC separations.
  • Rugged BEH particles offer superior chemical stability and extended column lifetimes.
  • Wide usable pH range addresses silica-based HILIC instability.
  • Packing materials are purpose-built for UPLC systems.
  • Manufactured in cGMP, ISO 9001 certified facilities with ultrapure reagents.
  • Each batch undergoes rigorous chromatographic testing.
  • Individually tested; Performance Chromatogram and Certificate of Batch Analysis included.