Learn More
Waters Corp ACQUITY UPLC BEH Phenyl Column, 130Å, 1.7 µm, 1 mm X 150 mm, 3/pk
Catalog No. 50780022 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This sorbent, along with the XBridge BEH Phenyl, provide you with the most chemiscally stable phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.
Catalog No. 50-780-022 Supplier Waters Corp Supplier No. 176001047
ACQUITY BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This sorbent, along with the XBridge BEH Phenyl, provide you with the most chemiscally stable phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.