Learn More
Waters Corp ACQUITY UPLC BEH Phenyl Method Validation Kit, 130Å, 1.7 µm, 2.1 mm X 50 mm, 3/pk
Catalog No. 50782156 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit consists of three ACQUITY UPLC BEH Phenyl columns packed with three different batches of sorbent.
Catalog No. 50-782-156 Supplier Waters Corp Supplier No. 186004050
This kit consists of three ACQUITY UPLC BEH Phenyl columns packed with three different batches of sorbent.