Learn More
Waters Corp WATERS CORPORATION Column, Phenyl, 1.7 μm, 2.1 mm x 75 mm
Supplier:  Waters Corp 186005607
Encompass_Preferred
ACQUITY UPLC BEH Phenyl columns offer complementary selectivity to C18 stationary phases, ideal for polyaromatic compounds. This sorbent provides high chemical stability, ultra-low column bleed, extended column lifetimes, and excellent peak shape.
  • Complementary selectivity to C18 for polyaromatic compounds.
  • Highly stable phenyl sorbent for robust performance.
  • Minimizes column bleed for cleaner baselines.
  • Extended operational lifetimes.
  • Superior peak shape.
  • Wide pH range of 1 to 12.
  • Withstands pressures up to 18000 psi (1240 Bar).
  • eCord™ technology for tracking usage.
Catalog No. 50-466-143