Learn More
Waters Corp ACQUITY UPLC Column, 130Å, 1.7 μm, 1 mm x 100 mm
Catalog No. 50781423 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY BEH Phenyl columns offer complementary selectivity for polyaromatic compounds. Designed for ultra-low bleed, long lifetimes, and excellent peak shape, they ensure chemical stability and repeatability. Manufactured under strict ISO 9001 standards, each column is tested for consistent performance with UPLC systems.
  • Complementary selectivity for polyaromatic compounds vs. C18 phases.
  • Ultra-low column bleed; long column lifetimes.
  • Excellent peak shape.
  • Trifunctionally linked phenyl-hexyl ligand for chemical stability and repeatability.
  • Manufactured to ISO 9001 standards using ultra-pure reagents.
  • Optimized for UPLC systems.
  • Each batch chromatographically tested.
  • Performance chromatogram and batch analysis on eCord™ Intelligent Chip.
  • Outstanding, repeatable performance guaranteed.
Catalog No. 50-781-423 Supplier Waters Corp Supplier No. 186002883
ACQUITY BEH Phenyl columns offer complementary selectivity for polyaromatic compounds. Designed for ultra-low bleed, long lifetimes, and excellent peak shape, they ensure chemical stability and repeatability. Manufactured under strict ISO 9001 standards, each column is tested for consistent performance with UPLC systems.
  • Complementary selectivity for polyaromatic compounds vs. C18 phases.
  • Ultra-low column bleed; long column lifetimes.
  • Excellent peak shape.
  • Trifunctionally linked phenyl-hexyl ligand for chemical stability and repeatability.
  • Manufactured to ISO 9001 standards using ultra-pure reagents.
  • Optimized for UPLC systems.
  • Each batch chromatographically tested.
  • Performance chromatogram and batch analysis on eCord™ Intelligent Chip.
  • Outstanding, repeatable performance guaranteed.