Learn More
Waters Corp Waters Corporation Column, Phenyl, 1.7 μm, 130 Å, 2.1 mm x 30 mm
Catalog No. 50782064 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY BEH Phenyl Column offers complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. This chemically stable phenyl sorbent provides ultra-low column bleed, long lifetimes, and excellent peak shape, ensuring high reproducibility and minimizing method variation.
  • Offers complementary selectivity to C18 stationary phases for polyaromatic compounds.
  • Chemically stable phenyl sorbent with ultra-low column bleed and long column lifetimes.
  • Ensures batch-to-batch reproducibility.
  • Features a wide pH stability range of 1 to 12.
  • Manufactured under strict ISO 9001 conditions.
  • Each column is individually tested; results available via eCord™ Intelligent Chip.
Catalog No. 50-782-064 Supplier Waters Corp Supplier No. 186003911
The ACQUITY BEH Phenyl Column offers complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. This chemically stable phenyl sorbent provides ultra-low column bleed, long lifetimes, and excellent peak shape, ensuring high reproducibility and minimizing method variation.
  • Offers complementary selectivity to C18 stationary phases for polyaromatic compounds.
  • Chemically stable phenyl sorbent with ultra-low column bleed and long column lifetimes.
  • Ensures batch-to-batch reproducibility.
  • Features a wide pH stability range of 1 to 12.
  • Manufactured under strict ISO 9001 conditions.
  • Each column is individually tested; results available via eCord™ Intelligent Chip.