Learn More
Waters Corp Waters Corporation ACQUITY UPLC BEH Phenyl Column, 1.7 μm, 130 Å, 3.0 × 75 mm
Catalog No. 50466148 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC BEH Phenyl columns are designed to integrate seamlessly with ACQUITY UPLC Systems. They feature sub-2-μm, fully porous particle technologies that deliver high efficiencies and the broadest sub-2-μm selectivity range. Utilizing rugged, base-particle technologies, these columns ensure best-in-class stability and offer maximum flexibility for high-throughput method development.
  • Sub-2-μm, fully porous particle technologies
  • High efficiencies
  • Broadest sub-2-μm selectivity range
  • Rugged, base-particle technologies
  • Best-in-class column stability
  • Maximum flexibility for high-throughput method development
  • Excellent peak asymmetry
  • Excellent efficiency and chemical stability
  • Robustness for operation under extreme pH conditions
  • Influences retention, selectivity, and sensitivity for ionizable compounds
Catalog No. 50-466-148 Supplier Waters Corp Supplier No. 186005612
ACQUITY UPLC BEH Phenyl columns are designed to integrate seamlessly with ACQUITY UPLC Systems. They feature sub-2-μm, fully porous particle technologies that deliver high efficiencies and the broadest sub-2-μm selectivity range. Utilizing rugged, base-particle technologies, these columns ensure best-in-class stability and offer maximum flexibility for high-throughput method development.
  • Sub-2-μm, fully porous particle technologies
  • High efficiencies
  • Broadest sub-2-μm selectivity range
  • Rugged, base-particle technologies
  • Best-in-class column stability
  • Maximum flexibility for high-throughput method development
  • Excellent peak asymmetry
  • Excellent efficiency and chemical stability
  • Robustness for operation under extreme pH conditions
  • Influences retention, selectivity, and sensitivity for ionizable compounds