Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters ACQUITY UPLC BEH Phenyl Column, 130Å, 1.7 μm, 2.1 mm x 150 mm

Catalog No. 50781727 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This column offers complementary selectivity to C18 stationary phases, designed for polyaromatic compounds. It provides high chemical stability, ultra-low column bleed, extended lifetimes, and excellent peak shapes for rapid, accurate analysis.

  • Complementary selectivity for polyaromatic compounds.
  • Highly stable phenyl sorbent; ultra-low column bleed.
  • Extended column lifetimes and excellent peak shapes.
  • Phenyl-hexyl ligand provides high stability and reproducibility.
  • Enables rapid analysis (up to 10x faster than HPLC).
  • Suitable for plastics, food, serum, and environmental samples.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Batch tested chromatographically for reproducible performance.
  • Particle size: 1.7 μm; Pore size: 130 Å.
  • Dimensions: 2.1 mm x 150 mm.
  • pH range: 1 to 12.
  • Max pressure: 18000 psi (1240 Bar).

Catalog No. 50-781-727 Supplier Waters Corp Supplier No. 186003378
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This column offers complementary selectivity to C18 stationary phases, designed for polyaromatic compounds. It provides high chemical stability, ultra-low column bleed, extended lifetimes, and excellent peak shapes for rapid, accurate analysis.

  • Complementary selectivity for polyaromatic compounds.
  • Highly stable phenyl sorbent; ultra-low column bleed.
  • Extended column lifetimes and excellent peak shapes.
  • Phenyl-hexyl ligand provides high stability and reproducibility.
  • Enables rapid analysis (up to 10x faster than HPLC).
  • Suitable for plastics, food, serum, and environmental samples.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Batch tested chromatographically for reproducible performance.
  • Particle size: 1.7 μm; Pore size: 130 Å.
  • Dimensions: 2.1 mm x 150 mm.
  • pH range: 1 to 12.
  • Max pressure: 18000 psi (1240 Bar).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.