Learn More
Waters Corp ACQUITY UPLC BEH Phenyl Column, 130Å, 1.7 μm
Catalog No. 50781724 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY BEH Phenyl columns provide complementary selectivity compared to C18 stationary phases, especially for polyaromatic compounds. This sorbent is designed to be highly chemically stable, ensuring ultra-low column bleed, extended column lifetimes, and excellent peak shape.
  • Offers complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds.
  • Features the most chemically stable phenyl sorbent available.
  • Results in ultra-low column bleed.
  • Provides long column lifetimes.
  • Delivers excellent peak shape.
Catalog No. 50-781-724 Supplier Waters Corp Supplier No. 186003375
ACQUITY BEH Phenyl columns provide complementary selectivity compared to C18 stationary phases, especially for polyaromatic compounds. This sorbent is designed to be highly chemically stable, ensuring ultra-low column bleed, extended column lifetimes, and excellent peak shape.
  • Offers complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds.
  • Features the most chemically stable phenyl sorbent available.
  • Results in ultra-low column bleed.
  • Provides long column lifetimes.
  • Delivers excellent peak shape.