Learn More
Waters Corp Waters ACQUITY UPLC Phenyl Column, 1.7 μm, 130 Å, 1 mm x 50 mm
Supplier:  Waters Corp 186002882
Encompass_Preferred
These columns provide complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. They offer a chemically stable phenyl sorbent, ensuring ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Chemically stable phenyl sorbent yields ultra-low column bleed and long column lifetimes.
  • Trifunctionally linked phenyl-hexyl ligand ensures high chemical stability and reproducibility.
  • Suitable for analytes with pore sizes of 130Å and 1.7 μm, with broad pH stability.
  • Industry-leading batch-to-batch reproducibility through ISO 9001 standards and stringent process control.
  • Each column is individually checked with a performance chromatogram and certificate of batch analysis.
Catalog No. 50-781-422