Learn More
Waters Corp Waters ACQUITY UPLC BEH Phenyl Column, 130Å, 1.7 μm, 2.1 mm x 150 mm
Catalog No. 50780026 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC columns are advanced LC columns, engineered for UPLC Technology pressures. They use highly efficient 1.7 μm Bridged Ethyl Hybrid (BEH) particles for maximum speed, sensitivity, and resolution. These Phenyl Columns enhance selectivity for aromatic analytes, offering high chemical stability, ultra-low column bleed, extended lifetime, and industry-leading pH stability.
  • Utilizes 1.7 μm Bridged Ethyl Hybrid (BEH) particles.
  • Provides maximum speed, sensitivity, and resolution.
  • Offers a wide pH range of 1 to 12.
  • Enhances selectivity for aromatic analytes.
  • Ensures high chemical stability, ultra-low column bleed, and extended lifetime.
  • Delivers excellent batch-to-batch reproducibility and outstanding peak shape.
  • Manufactured under ISO 9001 standards.
  • Features 2.1 mm ID x 150 mm length and 130 Å pore size.
  • Units per package: 3.
Catalog No. 50-780-026 Supplier Waters Corp Supplier No. 176001051
ACQUITY UPLC columns are advanced LC columns, engineered for UPLC Technology pressures. They use highly efficient 1.7 μm Bridged Ethyl Hybrid (BEH) particles for maximum speed, sensitivity, and resolution. These Phenyl Columns enhance selectivity for aromatic analytes, offering high chemical stability, ultra-low column bleed, extended lifetime, and industry-leading pH stability.
  • Utilizes 1.7 μm Bridged Ethyl Hybrid (BEH) particles.
  • Provides maximum speed, sensitivity, and resolution.
  • Offers a wide pH range of 1 to 12.
  • Enhances selectivity for aromatic analytes.
  • Ensures high chemical stability, ultra-low column bleed, and extended lifetime.
  • Delivers excellent batch-to-batch reproducibility and outstanding peak shape.
  • Manufactured under ISO 9001 standards.
  • Features 2.1 mm ID x 150 mm length and 130 Å pore size.
  • Units per package: 3.