Learn More
Waters Corp Waters ACQUITY UPLC BEH Phenyl Column, 130Å, 1.7 μm, 2.1 x 50 mm
Catalog No. 50781424 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns offer complementary selectivity to C18 stationary phases for polyaromatic compounds. They provide a chemically stable phenyl sorbent with ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Provides complementary selectivity to C18 stationary phases for polyaromatic compounds.
  • Features trifunctional C6-alkyl tether for enhanced stability.
  • Ensures ultra-low column bleed, long lifetimes, and excellent peak shape.
  • Utilizes 1.7 μm BEH particle technology for UPLC separations.
  • Improved column hardware minimizes band broadening and ensures leak-free connections.
  • Innovative frit design retains 1.7 μm particles.
  • Adheres to strict standards for reproducibility.
  • Operates within a pH range of 1 to 12.
  • Withstands operating pressures up to 18,000 psi (1240 Bar).
Catalog No. 50-781-424 Supplier Waters Corp Supplier No. 186002884
Add to Cart
Add to Cart
These columns offer complementary selectivity to C18 stationary phases for polyaromatic compounds. They provide a chemically stable phenyl sorbent with ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Provides complementary selectivity to C18 stationary phases for polyaromatic compounds.
  • Features trifunctional C6-alkyl tether for enhanced stability.
  • Ensures ultra-low column bleed, long lifetimes, and excellent peak shape.
  • Utilizes 1.7 μm BEH particle technology for UPLC separations.
  • Improved column hardware minimizes band broadening and ensures leak-free connections.
  • Innovative frit design retains 1.7 μm particles.
  • Adheres to strict standards for reproducibility.
  • Operates within a pH range of 1 to 12.
  • Withstands operating pressures up to 18,000 psi (1240 Bar).