Learn More
Waters Corp WATERS CORPORATION Column Kit, BEH Phenyl, 1.7 μm, 130 Å, Method Validation
Catalog No. 50466031 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit consists of three BEH Phenyl columns, each packed with a different batch of sorbent, designed for method validation.
  • Designed to streamline method validation processes by providing three columns with different sorbent batches.
  • Utilizes BEH Phenyl technology for efficient separations.
  • Features a wide pH range (1 to 12) and high maximum pressure (18000 psi (1240 Bar)) for versatile applications.
Catalog No. 50-466-031 Supplier Waters Corp Supplier No. 186004697
This kit consists of three BEH Phenyl columns, each packed with a different batch of sorbent, designed for method validation.
  • Designed to streamline method validation processes by providing three columns with different sorbent batches.
  • Utilizes BEH Phenyl technology for efficient separations.
  • Features a wide pH range (1 to 12) and high maximum pressure (18000 psi (1240 Bar)) for versatile applications.