Learn More
Waters Corp Waters Column, ACQUITY UPLC, BEH Shield RP18, 1.7 μm, 2.1 mm x 30 mm
Supplier:  Waters Corp 176001309
Encompass_Preferred
ACQUITY BEH Shield RP18 columns complement ACQUITY BEH C18 and C8 phases for HPLC method development. They feature an embedded carbamate group for alternative selectivity, especially for phenolic compounds, compared to traditional alkyl columns.
  • C18 chemistry, Reversed Phase separation mode
  • Hybrid particle substrate with spherical shape
  • pH range: 2 to 11
  • Maximum pressure: 18000 psi (1240 bar)
  • Endcapped; Low silanol activity
  • Shield RP18 bonding technology; BEH particle technology
  • 1.7 μm particle size, 130 Å pore size
  • Parker-style endfitting type
  • 185 surface area
  • Compatible with UPLC and UHPLC systems
  • L1 USP classification
  • 2.1 mm inner diameter, 30 mm length
  • 17% carbon load
  • eCord enabled
  • 3 units per package
Catalog No. 50-780-047