Learn More
Waters Corp ACQUITY UPLC Column, C18, 1.7 μm, 1 mm × 150 mm, 130 Å
Catalog No. 50781722 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is engineered to enhance HPLC method development, offering alternative selectivity for compounds, especially phenolics, due to its embedded carbamate group in the bonded phase ligand. It complements existing C18 and C8 phases.
  • C18 chemistry and reversed-phase separation mode
  • Hybrid particle substrate
  • pH range of 2 to 11
  • Maximum pressure of 18000 psi (1240 Bar)
  • Endcapped with Shield RP18 bonding technology
  • Low silanol activity
  • Spherical particle shape
  • 1.7 μm particle size
  • 130 Å pore size
  • Column format
  • Compatible with UPLC and UHPLC systems
  • BEH particle technology
  • L1 USP classification
  • 1 mm inner diameter
  • 150 mm length
  • 17% carbon load
  • eCord enabled
  • 1 unit per package
Catalog No. 50-781-722 Supplier Waters Corp Supplier No. 186003373
This column is engineered to enhance HPLC method development, offering alternative selectivity for compounds, especially phenolics, due to its embedded carbamate group in the bonded phase ligand. It complements existing C18 and C8 phases.
  • C18 chemistry and reversed-phase separation mode
  • Hybrid particle substrate
  • pH range of 2 to 11
  • Maximum pressure of 18000 psi (1240 Bar)
  • Endcapped with Shield RP18 bonding technology
  • Low silanol activity
  • Spherical particle shape
  • 1.7 μm particle size
  • 130 Å pore size
  • Column format
  • Compatible with UPLC and UHPLC systems
  • BEH particle technology
  • L1 USP classification
  • 1 mm inner diameter
  • 150 mm length
  • 17% carbon load
  • eCord enabled
  • 1 unit per package