Learn More
Waters Corp Waters Column, BEH Shield RP18, 1.7 μm, 2.1 x 30 mm
Catalog No. 50782062 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column offers alternate selectivity, providing superior peak asymmetry, efficiency, and chemical stability across a wide pH range.
  • Complements C18 and C8 phases for alternate selectivity.
  • Features an embedded carbamate group, enhancing selectivity for phenolic compounds.
  • C18 chemistry, reversed phase separation, and hybrid particle substrate.
  • Spherical 1.7 μm particles, 2.1 mm inner diameter, 30 mm length.
  • 130 Å pore size, 185 surface area, and 17 % carbon load.
  • Operates from pH 2 to 11 with maximum pressure of 18000 psi (1240 Bar).
  • Compatible with UPLC/UHPLC systems; utilizes BEH particle technology and eCord.
  • Delivers excellent peak asymmetry, efficiency, and chemical stability.
Catalog No. 50-782-062 Supplier Waters Corp Supplier No. 186003909
This column offers alternate selectivity, providing superior peak asymmetry, efficiency, and chemical stability across a wide pH range.
  • Complements C18 and C8 phases for alternate selectivity.
  • Features an embedded carbamate group, enhancing selectivity for phenolic compounds.
  • C18 chemistry, reversed phase separation, and hybrid particle substrate.
  • Spherical 1.7 μm particles, 2.1 mm inner diameter, 30 mm length.
  • 130 Å pore size, 185 surface area, and 17 % carbon load.
  • Operates from pH 2 to 11 with maximum pressure of 18000 psi (1240 Bar).
  • Compatible with UPLC/UHPLC systems; utilizes BEH particle technology and eCord.
  • Delivers excellent peak asymmetry, efficiency, and chemical stability.