Learn More
Waters Corp ACQUITY UPLC BEH Shield RP18 Column, 130Å, 1.7 µm, 3 mm X 50 mm, 1/pk
Catalog No. 50791177 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY BEH Shield RP18 columns complement the ACQUITY BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
Catalog No. 50-791-177 Supplier Waters Corp Supplier No. 186004668
ACQUITY BEH Shield RP18 columns complement the ACQUITY BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.