Learn More
Waters Corp Waters Column, BEH Shield RP18, 1.7 μm, 3 mm, 100 mm
Catalog No. 50791178 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed to complement BEH C18 and C8 phases, serving as a powerful tool for HPLC method development. They combine BEH technology with Shield Technology by integrating a carbamate into the bonded phase ligand, which provides alternate selectivity, particularly for phenolic compounds, when compared to straight-chain alkyl columns.
  • Enhanced C18 column selectivity for improved peak shape and high-resolution power.
  • Compatible with 100% aqueous mobile phases.
  • Embedded carbamate group provides alternative selectivity, especially for phenolic compounds.
  • Allows for operation within a broad pH range.
  • Utilizes the ultra-efficiency of the 1.7 μm BEH particle.
  • Combines the hydrophobicity of a straight-chain-alkyl C18 ligand with the hydrophilicity of an embedded carbamate polar group for outstanding results.
Catalog No. 50-791-178 Supplier Waters Corp Supplier No. 186004669
These columns are designed to complement BEH C18 and C8 phases, serving as a powerful tool for HPLC method development. They combine BEH technology with Shield Technology by integrating a carbamate into the bonded phase ligand, which provides alternate selectivity, particularly for phenolic compounds, when compared to straight-chain alkyl columns.
  • Enhanced C18 column selectivity for improved peak shape and high-resolution power.
  • Compatible with 100% aqueous mobile phases.
  • Embedded carbamate group provides alternative selectivity, especially for phenolic compounds.
  • Allows for operation within a broad pH range.
  • Utilizes the ultra-efficiency of the 1.7 μm BEH particle.
  • Combines the hydrophobicity of a straight-chain-alkyl C18 ligand with the hydrophilicity of an embedded carbamate polar group for outstanding results.