Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters SEC Column, 1.7 μm, 125 Å, 4.6 x 150 mm

Catalog No. 50201649 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This SEC column utilizes BEH particle technology, offering the first commercially available sub-2-μm SEC chemistry. Optimized for proteins and their aggregates (molecular weights from 1000 to 80,000 Daltons), it delivers fast, accurate aggregate determinations up to 10 times quicker than traditional HPLC methods. It provides exceptional improvements in resolution, throughput, and sensitivity for assays, enabling protein aggregate monitoring via size-exclusion chromatography (SEC).

  • Particle size: 1.7 μm
  • Pore size: 125 Å
  • Dimensions: 4.6 mm x 150 mm
  • Molecular weight range: 1000 to 80000 daltons
  • pH operating range: 1 to 12
  • Maximum pressure: 10000 psi (690 bar)
  • Compatible with UPLC and UHPLC systems
  • Faster aggregation species measurement (up to 10 times faster).
  • Unique DIOL coating minimizes undesired surface charges.

Catalog No. 50-201-649 Supplier Waters Corp Supplier No. 186006505
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This SEC column utilizes BEH particle technology, offering the first commercially available sub-2-μm SEC chemistry. Optimized for proteins and their aggregates (molecular weights from 1000 to 80,000 Daltons), it delivers fast, accurate aggregate determinations up to 10 times quicker than traditional HPLC methods. It provides exceptional improvements in resolution, throughput, and sensitivity for assays, enabling protein aggregate monitoring via size-exclusion chromatography (SEC).

  • Particle size: 1.7 μm
  • Pore size: 125 Å
  • Dimensions: 4.6 mm x 150 mm
  • Molecular weight range: 1000 to 80000 daltons
  • pH operating range: 1 to 12
  • Maximum pressure: 10000 psi (690 bar)
  • Compatible with UPLC and UHPLC systems
  • Faster aggregation species measurement (up to 10 times faster).
  • Unique DIOL coating minimizes undesired surface charges.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.