Learn More
Waters Corp Waters ACQUITY UPLC Peptide BEH C18 Column, 130Å, 1.7 μm, 2.1 mm x 100 mm
Catalog No. 50781838 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY BEH130 C18 Peptide Separation Technology (PST) columns are optimized for peptide separations. They feature a small-pore (130Å) trifunctionally bonded BEH particle, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed. This makes them suitable for proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Features a small-pore (130Å) trifunctionally bonded BEH particle.
  • Offers wide usable pH range, superior low pH stability, and ultra-low bleed.
  • Provides stable, reproducible peptide separations using BEH particle and C18 bonding.
  • Achieves ultimate resolution with sub-2 μm particles for diverse applications.
  • Ensures narrow, symmetrical peaks and good peak shape with retention in formic acid and TFA.
  • Stable at pH 1 to 12 and at elevated temperatures.
Catalog No. 50-781-838 Supplier Waters Corp Supplier No. 186003555
The ACQUITY BEH130 C18 Peptide Separation Technology (PST) columns are optimized for peptide separations. They feature a small-pore (130Å) trifunctionally bonded BEH particle, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed. This makes them suitable for proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Features a small-pore (130Å) trifunctionally bonded BEH particle.
  • Offers wide usable pH range, superior low pH stability, and ultra-low bleed.
  • Provides stable, reproducible peptide separations using BEH particle and C18 bonding.
  • Achieves ultimate resolution with sub-2 μm particles for diverse applications.
  • Ensures narrow, symmetrical peaks and good peak shape with retention in formic acid and TFA.
  • Stable at pH 1 to 12 and at elevated temperatures.