Learn More
Waters Corp ACQUITY UPLC Method Validation Kit, C18, 1.7 μm, 2.1 mm × 150 mm
Catalog No. 50201651 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. They feature a small-pore (130Å) trifunctionally bonded BEH particle, offering a wide usable pH range, excellent low pH stability, and minimal column bleed. This design is suitable for assaying samples in proteomics, protein characterization, and peptide synthesis.
  • Compatible with UPLC and UHPLC systems.
  • C18 chemistry, reversed phase separation mode, hybrid particle substrate.
  • Spherical particle shape with low silanol activity.
  • Molecular weight range of 1000 to 30000.
  • Parker-style endfitting type.
  • Carbon load of 18 %.
Catalog No. 50-201-651 Supplier Waters Corp Supplier No. 186006516
The ACQUITY BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. They feature a small-pore (130Å) trifunctionally bonded BEH particle, offering a wide usable pH range, excellent low pH stability, and minimal column bleed. This design is suitable for assaying samples in proteomics, protein characterization, and peptide synthesis.
  • Compatible with UPLC and UHPLC systems.
  • C18 chemistry, reversed phase separation mode, hybrid particle substrate.
  • Spherical particle shape with low silanol activity.
  • Molecular weight range of 1000 to 30000.
  • Parker-style endfitting type.
  • Carbon load of 18 %.