Learn More
Waters Corp Waters UPLC Column, C18, 300Å, 1.7 μm
Supplier:  Waters Corp 186005592
Encompass_Preferred
The ACQUITY BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. They feature wide-pore (300Å) trifunctionally bonded BEH particles, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed. These columns are designed for assaying samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Features wide-pore (300Å) trifunctionally bonded BEH particles.
  • Offers a broad usable pH range of 1 to 12.
  • Provides superior low pH stability and ultra-low column bleed.
  • Achieve narrow, symmetrical peaks for maximum resolution.
  • Manufactured in a cGMP, ISO 9002 certified plant.
  • Compatible with UPLC and PREMIER systems.
Catalog No. 50-466-135