Learn More
Waters Corp Waters ACQUITY UPLC Peptide BEH C18 Column, 1.7 μm, 300 Å, 1 mm x 100 mm
Supplier:  Waters Corp 186005593
Encompass_Preferred
The ACQUITY BEH300 C18 Peptide Separation Technology (PST) columns are optimized for peptide separations. They feature a wide-pore (300Å) trifunctionally bonded BEH particle, providing a broad usable pH range, superior low pH stability, and ultra-low column bleed, making them suitable for proteomics, protein characterization, and peptide synthesis.
  • Optimized and quality control tested for peptide separations.
  • Features wide-pore (300Å) trifunctionally bonded BEH particles, offering a broad usable pH range (1 to 12) and superior low pH stability.
  • Provides ultra-low column bleed for sensitive assays.
  • Achieves compact, symmetrical peaks for optimal resolution of diverse peptides.
  • Ensures reproducibility and consistency through stringent quality control validation with a peptide map.
  • Developed in a cGMP, ISO 9002 accredited plant and compatible with UPLC and UHPLC systems.
Catalog No. 50-466-136