Learn More
Waters Corp Waters BEH C18 Column, ACQUITY UPLC, 300Å, 1.7 µm
Catalog No. 50466137 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers a wide usable pH range, superior low pH stability, and ultra-low column bleed for analyzing samples in proteomics, protein characterization, and peptide synthesis.
  • Achieves narrow, symmetrical peaks for optimal resolution in peptide chromatography.
  • Separates a wide range of peptides: large, small, acidic, basic, hydrophobic, and hydrophilic.
  • Offers good peak shape and retention with formic acid and trifluoroacetic acid (TFA).
  • Features BEH Technology particles, QC certified with a peptide map for predictable separations.
  • Compatible with UPLC and PREMIER systems; manufactured in a cGMP, ISO 9002 certified plant.
Catalog No. 50-466-137 Supplier Waters Corp Supplier No. 186005594
The ACQUITY BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers a wide usable pH range, superior low pH stability, and ultra-low column bleed for analyzing samples in proteomics, protein characterization, and peptide synthesis.
  • Achieves narrow, symmetrical peaks for optimal resolution in peptide chromatography.
  • Separates a wide range of peptides: large, small, acidic, basic, hydrophobic, and hydrophilic.
  • Offers good peak shape and retention with formic acid and trifluoroacetic acid (TFA).
  • Features BEH Technology particles, QC certified with a peptide map for predictable separations.
  • Compatible with UPLC and PREMIER systems; manufactured in a cGMP, ISO 9002 certified plant.