Learn More
Waters Corp Waters ACQUITY UPLC Protein BEH C4 Column, 300 Å
Catalog No. 50782444 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC Protein BEH C4, 300 Å, 1.7 μm column is QC tested and optimized to separate proteins based on size, hydrophobicity, and isoelectric point. Compared to traditional C18 phases, the C4 ligand is less retentive, minimizing protein carryover, increasing protein recovery, and improving peak capacity.
  • QC tested and optimized for protein separation.
  • Separates proteins based on size, hydrophobicity, and isoelectric point.
  • C4 ligand provides less retention than C18, minimizing protein carryover and improving recovery and peak capacity.
  • Tailored for UPLC applications, offering sharp peak shapes and fast separations for proteins and peptides.
  • Ideal for separating medium to large proteins, such as monoclonal antibodies, and suitable for high throughput screening.
Catalog No. 50-782-444 Supplier Waters Corp Supplier No. 186004496
Add to Cart
Add to Cart
The ACQUITY UPLC Protein BEH C4, 300 Å, 1.7 μm column is QC tested and optimized to separate proteins based on size, hydrophobicity, and isoelectric point. Compared to traditional C18 phases, the C4 ligand is less retentive, minimizing protein carryover, increasing protein recovery, and improving peak capacity.
  • QC tested and optimized for protein separation.
  • Separates proteins based on size, hydrophobicity, and isoelectric point.
  • C4 ligand provides less retention than C18, minimizing protein carryover and improving recovery and peak capacity.
  • Tailored for UPLC applications, offering sharp peak shapes and fast separations for proteins and peptides.
  • Ideal for separating medium to large proteins, such as monoclonal antibodies, and suitable for high throughput screening.