Learn More
Waters Corp Waters Method Validation Kit, C4, 300 Å / 1.7 μm, 2.1 × 150 mm
Catalog No. 50466309 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three ACQUITY UPLC BEH300 C4 Protein Columns packed with three different batches of sorbent.
  • C4 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range from 1 to 10
  • Not endcapped
  • Low silanol activity
  • Maximum molecular weight range of 150,000
  • Spherical particle shape
  • 1.7 μm particle size
  • 300 Å pore size
  • 2.1 mm inner diameter
  • 150 mm length
  • 18% carbon load
  • Suitable for UPLC and UHPLC systems
  • BEH particle technology
  • L26 USP classification
  • 185 surface area
  • Protein QC tested
  • Designed for protein application
  • Parker-style endfitting type
  • 3 units per package
Catalog No. 50-466-309 Supplier Waters Corp Supplier No. 186006549
This method validation kit consists of three ACQUITY UPLC BEH300 C4 Protein Columns packed with three different batches of sorbent.
  • C4 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range from 1 to 10
  • Not endcapped
  • Low silanol activity
  • Maximum molecular weight range of 150,000
  • Spherical particle shape
  • 1.7 μm particle size
  • 300 Å pore size
  • 2.1 mm inner diameter
  • 150 mm length
  • 18% carbon load
  • Suitable for UPLC and UHPLC systems
  • BEH particle technology
  • L26 USP classification
  • 185 surface area
  • Protein QC tested
  • Designed for protein application
  • Parker-style endfitting type
  • 3 units per package