Learn More
Waters Corp Waters Column, C18, Peptide, 1.7 μm, 1 mm × 100 mm
Catalog No. 50596890 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Delivers excellent peak shape for peptides using FA- or TFA-containing mobile phases.
  • Exhibits little dependence on strong ion-pairing agents.
  • Ideal for LC or LC-MS applications.
  • Provides alternate selectivity for a wide range of chemicals.
  • Based on the Charged Surface Hybrid (CSH) particle platform.
  • Offers outstanding peak shape for basic chemicals.
  • Features enhanced loading capacity.
  • Designed for reduced column bleed.
  • Provides high efficiency.
  • Simulates TFA peak shape without using TFA.
  • Utilizes MS-friendly mobile phases like formic acid.
  • Enables faster and easier technique development.
  • Allows development of robust and transferable procedures.
  • Manufactured under strict conditions for consistent results.
  • Works well across a 1–11 pH range.
Catalog No. 50-596-890 Supplier Waters Corp Supplier No. 186006934
  • Delivers excellent peak shape for peptides using FA- or TFA-containing mobile phases.
  • Exhibits little dependence on strong ion-pairing agents.
  • Ideal for LC or LC-MS applications.
  • Provides alternate selectivity for a wide range of chemicals.
  • Based on the Charged Surface Hybrid (CSH) particle platform.
  • Offers outstanding peak shape for basic chemicals.
  • Features enhanced loading capacity.
  • Designed for reduced column bleed.
  • Provides high efficiency.
  • Simulates TFA peak shape without using TFA.
  • Utilizes MS-friendly mobile phases like formic acid.
  • Enables faster and easier technique development.
  • Allows development of robust and transferable procedures.
  • Manufactured under strict conditions for consistent results.
  • Works well across a 1–11 pH range.