Learn More
Waters Corp Waters Column, ACQUITY UPLC Peptide CSH C18, 1.7 μm, 1 mm x 50 mm, 130 Å
Catalog No. 50596889 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CSH130 C18 1.7 μm particles provide excellent peak shape for peptides using FA- or TFA-containing mobile phases, with little dependence on strong ion pairing agents. This makes the column suitable for LC or LC-MS applications and enables reliable, transferable procedures globally.
  • CSH technology mimics TFA peak shape with MS-friendly formic acid.
  • Enables faster technique development, avoiding ion-pairing agent effects.
  • Offers alternative selectivity for various chemicals.
  • Ensures outstanding peak shape and high loading capacity for basic chemicals at low pH.
  • Provides exceptional selectivity, minimal bleed, and high efficiency.
  • Manufactured under strict conditions for consistent results and long lifespan.
Catalog No. 50-596-889 Supplier Waters Corp Supplier No. 186006933
The CSH130 C18 1.7 μm particles provide excellent peak shape for peptides using FA- or TFA-containing mobile phases, with little dependence on strong ion pairing agents. This makes the column suitable for LC or LC-MS applications and enables reliable, transferable procedures globally.
  • CSH technology mimics TFA peak shape with MS-friendly formic acid.
  • Enables faster technique development, avoiding ion-pairing agent effects.
  • Offers alternative selectivity for various chemicals.
  • Ensures outstanding peak shape and high loading capacity for basic chemicals at low pH.
  • Provides exceptional selectivity, minimal bleed, and high efficiency.
  • Manufactured under strict conditions for consistent results and long lifespan.