Learn More
Waters Corp Waters Corporation C18 Column, 1.7 μm, 2.1 mm, 100 mm
Catalog No. 50596893 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters ACQUITY UPLC CSH130 C18 1.7 μm particles offer excellent peak shape for peptides with FA- or TFA-containing mobile phases. It's ideal for LC or LC-MS applications, showing little dependence on strong ion pairing agents.
  • Excellent peak shape for peptides (FA- or TFA mobile phases).
  • Low dependence on ion pairing agents; ideal for LC/LC-MS.
  • CSH particle platform; increased loading capacity.
  • Superior peak shape for basic compounds, low bleed, high efficiency.
  • Robust, transferable methods; TFA peak shape without TFA.
  • MS-friendly mobile phases (formic acid) for faster method development.
  • Tolerates pressures up to 18,000 psi (1241 bar / 124 MPa).
  • C18 chemistry, Reversed Phase, Hybrid substrate.
  • pH range: 1 to 11; Particle size: 1.7 μm; Pore size: 130 Å.
  • 2.1 mm ID, 100 mm Length; 15% Carbon load; Peptide application.
Catalog No. 50-596-893 Supplier Waters Corp Supplier No. 186006937
Waters ACQUITY UPLC CSH130 C18 1.7 μm particles offer excellent peak shape for peptides with FA- or TFA-containing mobile phases. It's ideal for LC or LC-MS applications, showing little dependence on strong ion pairing agents.
  • Excellent peak shape for peptides (FA- or TFA mobile phases).
  • Low dependence on ion pairing agents; ideal for LC/LC-MS.
  • CSH particle platform; increased loading capacity.
  • Superior peak shape for basic compounds, low bleed, high efficiency.
  • Robust, transferable methods; TFA peak shape without TFA.
  • MS-friendly mobile phases (formic acid) for faster method development.
  • Tolerates pressures up to 18,000 psi (1241 bar / 124 MPa).
  • C18 chemistry, Reversed Phase, Hybrid substrate.
  • pH range: 1 to 11; Particle size: 1.7 μm; Pore size: 130 Å.
  • 2.1 mm ID, 100 mm Length; 15% Carbon load; Peptide application.