Learn More
Waters Corp Waters VanGuard Pre-column, C18, 1.7 μm, 130 Å
Catalog No. 50596895 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 1.7 μm CSH130 C18 particles feature a low, carefully-defined positive charge concentration, ensuring excellent peak shape for peptides with FA- or TFA-containing mobile phases. This C18 column's performance shows little dependence on strong ion pairing agents, making it ideal for LC or LC-MS applications. The VanGuard pre-column connects directly to the inlet of an eXtended Performance column, minimizing extra column volumes and reducing leaking by eliminating additional connections.
  • Low, carefully-defined positive charges for excellent peptide peak shape with FA- or TFA-containing mobile phases.
  • Performance shows little dependence on strong ion pairing agents, ideal for LC or LC-MS applications.
  • Direct connection to the inlet of an eXtended performance column.
  • Minimizes extra column volumes.
  • Reduces leaking by eliminating additional connections.
Catalog No. 50-596-895 Supplier Waters Corp Supplier No. 186006939
These 1.7 μm CSH130 C18 particles feature a low, carefully-defined positive charge concentration, ensuring excellent peak shape for peptides with FA- or TFA-containing mobile phases. This C18 column's performance shows little dependence on strong ion pairing agents, making it ideal for LC or LC-MS applications. The VanGuard pre-column connects directly to the inlet of an eXtended Performance column, minimizing extra column volumes and reducing leaking by eliminating additional connections.
  • Low, carefully-defined positive charges for excellent peptide peak shape with FA- or TFA-containing mobile phases.
  • Performance shows little dependence on strong ion pairing agents, ideal for LC or LC-MS applications.
  • Direct connection to the inlet of an eXtended performance column.
  • Minimizes extra column volumes.
  • Reduces leaking by eliminating additional connections.