Learn More
Waters Corp Waters Column, C18, 1.7 μm, 3 mm x 75 mm, 130 Å
Supplier:  Waters Corp 186005623
Encompass_Preferred
ACQUITY CSH C18 columns offer alternative selectivity compared to other reversed-phase UPLC columns. They are designed to overcome issues such as peak shape asymmetry, poor loading capacity for basic compounds, column bleed, and rapid mobile-phase re-equilibration. The column utilizes Charged Surface Hybrid (CSH) Technology for quicker and simpler method development.
  • Provides Trifluoroacetic Acid (TFA) peak shape performance without the need for TFA.
  • Produces better peak shape performance with MS-friendly mobile phase additives like formic acid.
  • Offers a wide pH range of 1 to 11.
  • Sustains a maximum pressure of 18,000 psi (1240 Bar).
  • Allows seamless transferability between different particle sizes and column formats.
  • Offers alternative selectivity and a unique design without sacrificing essential performance characteristics.
Catalog No. 50-466-159