Learn More
Waters Corp Waters ACQUITY UPLC Fluoro-Phenyl Column, 130Å, 1.7 μm, 3 x 100 mm
Catalog No. 502037702 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY CSH Fluoro-Phenyl column is designed to maximize selectivity differences and provides maximum method development freedom, particularly when using low pH mobile phases. Its non-encapped sorbent offers enhanced retention of acidic compounds.
  • Designed to maximize selectivity differences.
  • Provides maximum method development freedom.
  • Enhanced retention of acidic compounds due to non-encapped sorbent.
Catalog No. 50-203-7702 Supplier Waters Corp Supplier No. 186005356
The ACQUITY CSH Fluoro-Phenyl column is designed to maximize selectivity differences and provides maximum method development freedom, particularly when using low pH mobile phases. Its non-encapped sorbent offers enhanced retention of acidic compounds.
  • Designed to maximize selectivity differences.
  • Provides maximum method development freedom.
  • Enhanced retention of acidic compounds due to non-encapped sorbent.