Learn More
Waters Corp ACQUITY UPLC Flex Cart, Mobile, Height-Adjustable, 29 to 41.5 in.
Catalog No. 50791713 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC Flex Cart is an optional component designed for the ACQUITY UPLC System with Column Manager. It provides a mobile and height-adjustable platform for the system.
  • Adjustable height allows for a customizable working height from 29 in. to 41.5 in.
  • Mobile design provides ease of movement for the UPLC system.
  • Robust support capable of supporting up to 400 lb, providing a stable platform for system components.
  • Integrates as an optional component with the ACQUITY UPLC System with Column Manager.
Catalog No. 50-791-713 Supplier Waters Corp Supplier No. 205015015
Add to Cart
Add to Cart
Request a Quote
The ACQUITY UPLC Flex Cart is an optional component designed for the ACQUITY UPLC System with Column Manager. It provides a mobile and height-adjustable platform for the system.
  • Adjustable height allows for a customizable working height from 29 in. to 41.5 in.
  • Mobile design provides ease of movement for the UPLC system.
  • Robust support capable of supporting up to 400 lb, providing a stable platform for system components.
  • Integrates as an optional component with the ACQUITY UPLC System with Column Manager.