Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp ACQUITY UPLC Glycoprotein Column, 300Å, 1.7 μm

Catalog No. 502390544 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This kit features columns engineered for the analysis of intact glycoproteins, glycoprotein fragments, and glycopeptides. Each column undergoes rigorous quality control testing with a Glycoprotein Performance Test Standard, ensuring consistent performance across batches. The kit includes the columns and a vial of the Glycoprotein Performance Test Standard, enabling immediate performance benchmarking upon receipt.

  • Chemistry: Amide
  • Separation mode: Hydrophilic interaction (HILIC)
  • Particle substrate: Hybrid
  • Particle size: 1.7 μm
  • Pore size: 300 Å
  • Inner diameter: 2.1 mm
  • Length: 100 mm
  • Units per package: 3 pk
  • pH range: 2 to 10
  • Temperature range: 20°C to 90°C
  • Pressure tolerance: Up to 15,000 psi (1034 bar or 103 MPa)
  • High resolving power
  • Outstanding chemical and mechanical stability

Catalog No. 50-239-0544 Supplier Waters Corp Supplier No. 176003703
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit features columns engineered for the analysis of intact glycoproteins, glycoprotein fragments, and glycopeptides. Each column undergoes rigorous quality control testing with a Glycoprotein Performance Test Standard, ensuring consistent performance across batches. The kit includes the columns and a vial of the Glycoprotein Performance Test Standard, enabling immediate performance benchmarking upon receipt.

  • Chemistry: Amide
  • Separation mode: Hydrophilic interaction (HILIC)
  • Particle substrate: Hybrid
  • Particle size: 1.7 μm
  • Pore size: 300 Å
  • Inner diameter: 2.1 mm
  • Length: 100 mm
  • Units per package: 3 pk
  • pH range: 2 to 10
  • Temperature range: 20°C to 90°C
  • Pressure tolerance: Up to 15,000 psi (1034 bar or 103 MPa)
  • High resolving power
  • Outstanding chemical and mechanical stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.