Learn More
Waters Corp Waters UPLC Column, Amide, 300Å, 1.7 μm, 2.1 mm x 100 mm
Supplier: Waters Corp 186007962
These Glycoprotein BEH Amide 300Å, 1.7μm columns are primarily designed for the analysis of intact glycoproteins, glycoprotein fragments, and glycopeptides. Each batch of Glycoprotein BEH Amide 300Å 1.7μm material undergoes specific quality control testing with the Glycoprotein Performance Test Standard to ensure batch-to-batch consistency.
- Chemistry: Amide
- Separation mode: Hydrophilic Interaction (HILIC)
- Particle substrate: Hybrid
- pH range: 2 to 10
- Particle size: 1.7 μm
- Pore size: 300 Å
- Format: Column
- System: UPLC, UHPLC
- Technique: LC
- Inner diameter: 2.1 mm
- Length: 100 mm
- Units per package: 1 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.