Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp ACQUITY UPLC VanGuard Pre-column, Amide, 300Å, 1.7 μm, 2.1 mm × 5 mm

Catalog No. 502037433 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These columns analyze intact glycoproteins, fragments, and glycopeptides. Each batch undergoes quality control with a performance test standard, including Ribonuclease A and B glycoforms, ensuring batch-to-batch consistency. The kit contains one column and a vial of the standard for immediate benchmarking. The standard can be reordered for continuous monitoring, with each batch prepared under strict quality control, confirmed by a certificate of analysis.

  • Amide chemistry
  • Hydrophilic interaction (HILIC) separation mode
  • Hybrid particle substrate
  • pH range 2 to 10
  • Maximum pressure 18000 psi (1240 bar)
  • Not endcapped
  • Particle size 1.7 μm
  • Pore size 300 Å
  • QC tested for glycoprotein
  • VanGuard pre-column
  • Compatible with UPLC/UHPLC
  • BEH particle technology
  • Inner diameter 2.1 mm
  • Length 5 mm
  • 3 units per package

Catalog No. 50-203-7433 Supplier Waters Corp Supplier No. 176003699
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These columns analyze intact glycoproteins, fragments, and glycopeptides. Each batch undergoes quality control with a performance test standard, including Ribonuclease A and B glycoforms, ensuring batch-to-batch consistency. The kit contains one column and a vial of the standard for immediate benchmarking. The standard can be reordered for continuous monitoring, with each batch prepared under strict quality control, confirmed by a certificate of analysis.

  • Amide chemistry
  • Hydrophilic interaction (HILIC) separation mode
  • Hybrid particle substrate
  • pH range 2 to 10
  • Maximum pressure 18000 psi (1240 bar)
  • Not endcapped
  • Particle size 1.7 μm
  • Pore size 300 Å
  • QC tested for glycoprotein
  • VanGuard pre-column
  • Compatible with UPLC/UHPLC
  • BEH particle technology
  • Inner diameter 2.1 mm
  • Length 5 mm
  • 3 units per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.