Learn More
Waters Corp ACQUITY UPLC Column, HSS C18, 100Å, 1.8 μm, 2.1 mm x 30 mm
Catalog No. 50780052 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HSS C18 column features ultra-performance C18 bonded phase chemistry, providing superior peak shape for basic compounds and increased retention. It is designed for extended column lifetimes, even under low pH conditions, and is fully scalable and transferable to HPLC separations.
  • C18 chemistry for superior peak shape, increased retention, and long column lifetime at low pH.
  • Reversed phase separation on silica, pH range 1 to 8, 45°C temperature limit.
  • Max pressure 18000 psi (1240 Bar); endcapped, low silanol, spherical 1.8 μm particles.
  • 100 Å pore size; HSS particle technology for UPLC/UHPLC.
  • 2.1 mm ID, 30 mm length, 15% carbon load, eCord enabled.
  • Fully scalable/transferable to HPLC; supplied in a 3-unit package.
Catalog No. 50-780-052 Supplier Waters Corp Supplier No. 176001398
This HSS C18 column features ultra-performance C18 bonded phase chemistry, providing superior peak shape for basic compounds and increased retention. It is designed for extended column lifetimes, even under low pH conditions, and is fully scalable and transferable to HPLC separations.
  • C18 chemistry for superior peak shape, increased retention, and long column lifetime at low pH.
  • Reversed phase separation on silica, pH range 1 to 8, 45°C temperature limit.
  • Max pressure 18000 psi (1240 Bar); endcapped, low silanol, spherical 1.8 μm particles.
  • 100 Å pore size; HSS particle technology for UPLC/UHPLC.
  • 2.1 mm ID, 30 mm length, 15% carbon load, eCord enabled.
  • Fully scalable/transferable to HPLC; supplied in a 3-unit package.