Learn More
Waters Corp Waters ACQUITY UPLC HSS C18 Column, 1.8 μm, 100 Å, 1 mm × 50 mm
Catalog No. 50781819 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase designed to provide superior peak shape for bases, increased retention compared to ACQUITY UPLC BEH C18 columns, and exceptionally long column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations.
  • Superior peak shape for bases
  • Increased retention
  • Extremely long column lifetimes at low pH
  • Fully scalable and transferable to HPLC separations
Catalog No. 50-781-819 Supplier Waters Corp Supplier No. 186003529
ACQUITY UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase designed to provide superior peak shape for bases, increased retention compared to ACQUITY UPLC BEH C18 columns, and exceptionally long column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations.
  • Superior peak shape for bases
  • Increased retention
  • Extremely long column lifetimes at low pH
  • Fully scalable and transferable to HPLC separations