Learn More
Waters Corp Waters Column, C18, 1.8 μm, 2.1 mm × 100 mm, 100 Å
Catalog No. 50780037 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase, designed for superior peak shape for bases, increased retention, and long column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations. They utilize High Strength Silica (HSS) technology, built to withstand UPLC pressures.
  • Superior peak shape for basic compounds.
  • Increased retention compared to hybrid particle technology.
  • Extended column lifetimes at low pH.
  • Scalable and transferable to HPLC separations.
  • HSS technology provides mechanical stability under UPLC pressures.
  • Excellent peak shapes and low pH stability.
  • Facilitates low pH separations of complex mixtures.
  • Low column bleed and excellent batch-to-batch reproducibility.
  • Recommended operating temperature of 20–45 °C.
Catalog No. 50-780-037 Supplier Waters Corp Supplier No. 176001125
This UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase, designed for superior peak shape for bases, increased retention, and long column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations. They utilize High Strength Silica (HSS) technology, built to withstand UPLC pressures.
  • Superior peak shape for basic compounds.
  • Increased retention compared to hybrid particle technology.
  • Extended column lifetimes at low pH.
  • Scalable and transferable to HPLC separations.
  • HSS technology provides mechanical stability under UPLC pressures.
  • Excellent peak shapes and low pH stability.
  • Facilitates low pH separations of complex mixtures.
  • Low column bleed and excellent batch-to-batch reproducibility.
  • Recommended operating temperature of 20–45 °C.