Learn More
Waters Corp Waters Column, C18, 100Å, 1.8 μm, 2.1 mm X 100 mm
Catalog No. 50781823 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC HSS C18 chemistry is a general-purpose C18 bonded phase designed for ultra-performance liquid chromatography. It provides superior peak shape for bases, increased retention, and extended column lifetimes at low pH, making it scalable and transferable to HPLC separations. This column is ideal for high-efficiency separations of small molecules in pharmaceutical, environmental, and food safety applications.
  • General-purpose C18 column with excellent peak shapes and low-pH stability.
  • Increased retention compared to hybrid-based C18 columns.
  • HSS (High Strength Silica) particle technology withstands UPLC pressures.
  • Ideal for high-efficiency separations in pharmaceutical, environmental, and food safety applications.
  • Supports a broad temperature range (up to 80°C) for enhanced analyte diffusion and reduced run times.
  • Compatible with UPLC and UHPLC systems.
Catalog No. 50-781-823 Supplier Waters Corp Supplier No. 186003533
The ACQUITY UPLC HSS C18 chemistry is a general-purpose C18 bonded phase designed for ultra-performance liquid chromatography. It provides superior peak shape for bases, increased retention, and extended column lifetimes at low pH, making it scalable and transferable to HPLC separations. This column is ideal for high-efficiency separations of small molecules in pharmaceutical, environmental, and food safety applications.
  • General-purpose C18 column with excellent peak shapes and low-pH stability.
  • Increased retention compared to hybrid-based C18 columns.
  • HSS (High Strength Silica) particle technology withstands UPLC pressures.
  • Ideal for high-efficiency separations in pharmaceutical, environmental, and food safety applications.
  • Supports a broad temperature range (up to 80°C) for enhanced analyte diffusion and reduced run times.
  • Compatible with UPLC and UHPLC systems.