Learn More
Waters Corp Waters ACQUITY UPLC HSS C18 Column, 100Å, 1.8μm, 1 mm x 50 mm
Supplier:  Waters Corp 176001121
Encompass_Preferred
This HSS C18 chemistry is a general-purpose C18 bonded phase designed for ultra-performance liquid chromatography. It offers superior peak shape for bases, increased retention, and extended column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations.
  • Provides superior peak shape for bases.
  • Offers increased retention for both polar and non-polar analytes.
  • Delivers extended column lifetimes at low pH.
  • Fully scalable and transferable to HPLC separations.
  • Utilizes a thermally treated silica particle for mechanical strength and stability under UPLC system pressures.
  • Maximizes selectivity space, enabling traditional hydrophobic, reversed-phase, dipole-dipole, aromatic, and hydrogen-bonding interactions.
Catalog No. 50-780-033