Learn More
Waters Corp Waters ACQUITY UPLC HSS C18 Column, 100 Å, 1.8 μm, 2.1 mm × 50 mm
Catalog No. 50780036 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase that offers superior peak shape for bases, increased retention compared to BEH C18 columns, and extremely long column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations.
  • Ultra-performance general purpose C18 bonded phase.
  • Provides superior peak shape for bases.
  • Increased retention compared to BEH C18 columns.
  • Extremely long column lifetimes at low pH.
  • Fully scalable and transferable to HPLC separations.
  • Designed to give reliable and reproducible results.
  • Manufactured in a cGMP-certified, ISO 9001-certified facility using ultra-pure reagents.
  • Each batch of packing material is chromatographically tested with acidic, basic, and neutral analytes.
Catalog No. 50-780-036 Supplier Waters Corp Supplier No. 176001124
Add to Cart
Add to Cart
The ACQUITY UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase that offers superior peak shape for bases, increased retention compared to BEH C18 columns, and extremely long column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations.
  • Ultra-performance general purpose C18 bonded phase.
  • Provides superior peak shape for bases.
  • Increased retention compared to BEH C18 columns.
  • Extremely long column lifetimes at low pH.
  • Fully scalable and transferable to HPLC separations.
  • Designed to give reliable and reproducible results.
  • Manufactured in a cGMP-certified, ISO 9001-certified facility using ultra-pure reagents.
  • Each batch of packing material is chromatographically tested with acidic, basic, and neutral analytes.