Learn More
Waters Corp Waters ACQUITY UPLC HSS C18 Column, 100Å, 1.8 μm, 1 mm x 100 mm, 3/pk
Supplier:  Waters Corp 176001122
Encompass_Preferred
The ACQUITY UPLC HSS C18 chemistry is designed as an ultra-performance general-purpose C18 bonded phase. It is engineered to deliver superior peak shape for bases and offers increased retention compared to ACQUITY UPLC BEH C18 columns. These columns are known for their extremely long lifetimes, particularly when operating at low pH. Additionally, ACQUITY HSS C18 columns are fully scalable and transferable, allowing for seamless integration with HPLC separations.
  • Ultra-performance general purpose C18 bonded phase.
  • Provides superior peak shape for bases.
  • Increased retention (compared to BEH C18 columns).
  • Extremely long column lifetimes at low pH.
  • Fully scalable and transferable to HPLC separations.
Catalog No. 50-780-034